Key Documents
Z516694
Sigma-Aldrich
Aldrich® three-neck round-bottom flask
vertical, capacity 500 mL, joint: ST/NS 34/45, side Joints: ST/NS 24/40 (2)
Synonym(s):
flask round bottom, round bottom flasks, round bottomed flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
three-neck (Vertical)
three-neck
joint
side Joints: ST/NS24/40 (2)
joint: ST/NS34/45
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)