Key Documents
Z514640
Sigma-Aldrich
Aldrich® three-neck round-bottom flask
vertical, capacity 500 mL, joint: ST/NS 29/42
Synonym(s):
flask round bottom, round bottom flasks, round bottomed flask
About This Item
UNSPSC Code:
41121804
material
glass
round bottom flask
three-neck (Vertical)
joint
joint: ST/NS29/42
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)