Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z512338

Hirschmann® Solarus® solar-powered manual titrator

volume 20 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

incremental 0.01 mL
with recirculating valve

manufacturer/tradename

Hirschmann 9392020

volume

20 mL

fitting

GL 32
GL 38
S 40

Looking for similar products? Visit Product Comparison Guide

General description

Integrated solar panels supply all of the energy for the electronic components and display, even under limited lighting conditions, i.e. no more charging or changing batteries. Designed to replace glass burettes and expensive electronic titrators. Fits Pyrex® and Duran® media bottles with GL 45 threads, and includes adapters to fit Aldrich Sealed for Quality and Sure/Seal bottles (GL 32), 1 L and gal. bottles (GL 38), and bottles with S40 threads.
  • Easily readable, user-friendly LC readout
  • Precise hand wheels with soft touch surfaces
  • Reagent recirculation system
  • Serial, bidirectional RS 232 interface to record data
  • Rotates 360° on bottle for easy readings
  • Quick-Cal for user specific calibration or use default
Specifications:
Increment:10 μL
Volume accuracy:Absolute (R) 0.2%; standard deviation (CV) 0.1%
Piston:ECTFE, PTFE
Cylinder: Borosilicate glass
Valves:Ball and seat borosilicate glass, Hastelloy® valve spring
Tubes:Suction and dispensing tubes are FEP
Sterilization:All components in contact with media are steam sterilizable
Certification:Individually tested, certified, and serial numbered
Connections:Luer lock connection for filters and drying tubes
Power:Solar panels

Legal Information

DURAN is a registered trademark of DWK Life Sciences
Hastelloy is a registered trademark of Haynes International, Inc.
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
PYREX is a registered trademark of Corning, Inc.
Solarus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service