Key Documents
Z511714
Microtip probe
tip diam. 13 mm, volume range 50-150 mL
About This Item
UNSPSC Code:
42281712
NACRES:
NB.74
material
colorless tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 1
manufacturer/tradename
Sonics & Materials 630-0561
tip diam.
13 mm
volume range
50-150 mL
General description
For processing small samples at extremely high-intensity. Probes can reach down into Eppendorf tubes, culture tubes, scintillation vials, etc. Made of titanium, autoclavable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)