Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z511161

Sigma-Aldrich

Solid-phase peptide synthesis flask

capacity 5,000 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.24

sterility

non-sterile

feature

closure type Cap

packaging

pack of 1

manufacturer/tradename

Sigma-Aldrich

capacity

5,000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Integral stirrer allows for the complete suspension of media.
Large 85 mm top and bottom openings allow for easy access when loading and cleaning.

Complete assembly includes top cap, funnel, coarse frit, stirrer assembly, 90 mm base assembly, 3 ft. of 1/4 in. PTFE tubing, 1/4 in. nut and ferrule. (Two additional ports are provided for reactant addition or gas inlet).

Features include: A removable fritted disc and a large bore 1/4-turn bottom valve for rapid filtration and washing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service