Z511153
Solid-phase peptide synthesis flask
capacity 2,000 mL
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.24
Recommended Products
sterility
non-sterile
feature
closure type Cap
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
2,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Integral stirrer allows for the complete suspension of media.
Large 85 mm top and bottom openings allow for easy access when loading and cleaning.
Complete assembly includes top cap, funnel, coarse frit, stirrer assembly, 90 mm base assembly, 3 ft. of 1/4 in. PTFE tubing, 1/4 in. nut and ferrule. (Two additional ports are provided for reactant addition or gas inlet).
Features include: A removable fritted disc and a large bore 1/4-turn bottom valve for rapid filtration and washing.
Large 85 mm top and bottom openings allow for easy access when loading and cleaning.
Complete assembly includes top cap, funnel, coarse frit, stirrer assembly, 90 mm base assembly, 3 ft. of 1/4 in. PTFE tubing, 1/4 in. nut and ferrule. (Two additional ports are provided for reactant addition or gas inlet).
Features include: A removable fritted disc and a large bore 1/4-turn bottom valve for rapid filtration and washing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service