Z510297
Aldrich® high-performance System 45™ glass kit
with threaded PBT/PTFE adapters
About This Item
Recommended Products
General description
A simple twist of the threaded PBT/PTFE adapters effects a grease-free, gas-tight seal at all glass connections. Flasks have wide, easy-access GL 25 threaded openings for excellent sample recovery and for thorough cleaning. The addition funnel features a precision control valve with an unbreakable PTFE tip. Quick-disconnects are utilized at all tubing connections. Leak-free PTFE-lined screw caps replace glass stoppers in this system.
Legal Information
Kit Components Only
- Vacuum-distilling adapter, joint: ST/NS 14/20, with quick-disconnect
- Septum-inlet cap, Septum-inlet cap
- Replacement cap liners, PTFE/silicone
Kit Components Also Available Separately
- Z510351Vigreux distilling column, bottom joint: ST/NS 14/20SDS
- Z510424Round-bottom flask, capacity 100 mLSDS
- Z510416Round-bottom flask, capacity 50 mLSDS
- Z510408Round-bottom flask, capacity 25 mLSDS
- Z510335Connecting adapter, bottom joint: ST/NS 14/20, side joint: ST/NS 14/20SDS
- Z510300Drying tube, joint: ST/NS 14/20SDS
- Z510343Claisen adapter, bottom joint: ST/NS 14/20SDS
- Z510319Vacuum adapter, joint: ST/NS 14/20, with quick-disconnectSDS
- Z510378Liebig condenser, joint: ST/NS 14/20, with two quick-disconnectsSDS
- Z510386Dropping funnel, capacity 50 mL, with PTFE valveSDS
- Z510440Gas-inlet tube/thermometer adapter, pkg of 1 eaSDS
- Z510432Round-bottom flask, capacity 100 mL, 3-neck, angledSDS
- Z510661Joint adapter, joint: ST/NS 14/20, PTFE (PBT)SDS
- Z512257Aldrich® System 45™ solid-top caps, diam. 25 mmSDS
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service