Key Documents
Z510203
Sigma-Aldrich
Large-capacity Kugelrohr glassware sets with System 45 compatible connections
2-L glassware set
About This Item
UNSPSC Code:
41104812
General description
Sets contain:
1 – Straight tube with removable in. 1/4 in. I.D. hose connection
1 – Single-bulb ball-tube flask
1 – Round-bottom oven flask
2 – PBT/PTFE screw-thread connectors
3 – Solid-top caps, PBT with PTFE liner
PBT/PTFE connectors resist heat to 200 °C and may be used interchangeably with 1- and 2-liter glassware. Special "reverse threading" on these flasks prevents connections from loosening during rotation.
Legal Information
System 45 is a trademark of John Babashak
Choose from one of the most recent versions:
Certificates of Analysis (COA)