Key Documents
Z509663
Sigma-Aldrich
Test papers
Neutral Litmus
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
feature
non-bleeding: no
visual transition interval
4.5-8.3, red to blue
detection method
colorimetric
General description
Package contains 100 strips and match the color of the paper inside for easy identification Strips are 2 L × 1/4 in. W.
Choose from one of the most recent versions:
Certificates of Analysis (COA)