Key Documents
Z509531
Support ring
ring diam. 7 in.
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
ring diam.
7 in.
General description
Cast iron ring with integral support clamp to attach to standard supports. Fits 1/2 in. diameter rods.
Choose from one of the most recent versions:
Certificates of Analysis (COA)