Key Documents
Z508241
Quick-release clamps
with extension, for flange 100 mm
Synonym(s):
clamps, quick release clamp
About This Item
UNSPSC Code:
41122413
NACRES:
NB.43
manufacturer/tradename
Ace Glass 651754
General description
SS clamps for use with reaction flasks and heads with a 100 mm Schott flange. The extension rod can be clamped to the support frame and helps stabilize the reaction assembly.
Choose from one of the most recent versions:
Certificates of Analysis (COA)