Key Documents
Z507148
Supelco
Michel-Miller Swagelok® adapter
NPTF 1/8 in., Nylon, Ace-Thred for #7
About This Item
UNSPSC Code:
12000000
material
Nylon
manufacturer/tradename
Ace Glass 584416
technique(s)
LPLC: suitable
NPTF
1/8 in.
fitting
Ace-Thred for #7
General description
Use to make Swagelok tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with #50 Ace-Thred fit Ace large columns.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)