Key Documents
Z507105
Supelco
Michel-Miller column plug
Front seal, PTFE, thread size 15 mm
About This Item
UNSPSC Code:
12000000
material
PTFE
manufacturer/tradename
Ace Glass 584648
technique(s)
LPLC: suitable
thread size
15 mm
General description
Solid plug for use as stopper with Ace-Threds. Seals M-M column ends, Plugs with #50 Ace-Thred fit Ace large columns.
Choose from one of the most recent versions:
Certificates of Analysis (COA)