Key Documents
Z507032
Supelco
Michel-Miller column plug
Front seal, Nylon, thread size 11 mm
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
material
Nylon
manufacturer/tradename
Ace Glass 584606
technique(s)
LPLC: suitable
thread size
11 mm
General description
Solid plug for use as stopper with Ace-Threds. Seals M-M column ends, Plugs with #50 Ace-Thred fit Ace large columns.
Choose from one of the most recent versions:
Certificates of Analysis (COA)