Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z423408

Easy-Read® thermometer

temperature -20-110 °C, immersion level, total

Synonym(s):

immersion thermometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303
NACRES:
NB.79

feature

immersion level total

manufacturer/tradename

HB Instruments B60304-0100

temperature measuring range

-20-110 °C

L

305 mm

O.D.

6-7 mm

Looking for similar products? Visit Product Comparison Guide

General description

Easy-to-read thermometers with black printing on a bright yellow background. Filled with contrasting black, non-hazardous, Enviro-Safe liquid. Calibrated with NIST, DKD standards.

Application

Easy Read® thermometer can be used in pharmaceutical, petrochemical, food, beverage processing, dairies, breweries, distilleries, universities and chemical labs.

Legal Information

Easy Read is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service