Key Documents
Z417033
Sigma-Aldrich
Aldrich® System 45 open-top caps
diam. 32 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
packaging
pkg of 10
diam.
32 mm
General description
Caps are PBT. Temp. range: -45 to 180°C. For use with PTFE-faced silicone liners listed below. Use with syringes for reagent introduction or sampling.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
Choose from one of the most recent versions:
Certificates of Analysis (COA)