Key Documents
Z416401
Supelco
EZSafe® flash-chromatography assembly
capacity 600 mL, With fritted disc (porosity C)
About This Item
UNSPSC Code:
12352200
NACRES:
NC.05
technique(s)
LPLC: suitable
capacity
600 mL
suitability
suitable for chromatography
General description
These columns are improved low-pressure columns (max. pressure 7psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy.
  • Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE
  • GL-45 screw-thread connections ensure positive reliable connections
  • Eliminates frozen PTFE and glass plugs
  • Eliminates grease
  • Choose between standard columns or columns with a fritted disc
The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.
Legal Information
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)