Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z416274

Supelco

EZSafe® flash-chromatography assembly

capacity 600 mL, Standard column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

for chromatography

technique(s)

LPLC: suitable

capacity

600 mL

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

General description

These columns are improved low-pressure columns (max. pressure 7psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy.
  • Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE
  • GL-45 screw-thread connections ensure positive reliable connections
  • Eliminates frozen PTFE and glass plugs
  • Eliminates grease
  • Choose between standard columns or columns with a fritted disc

The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.

Legal Information

EZ Safe is a registered trademark of Gerresheimer Glass, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service