Key Documents
Z414360
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock on side-arm
joint: ST/NS 24/40, PTFE stopcock
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
PTFE stopcock
joint
joint: ST/NS24/40
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)