Key Documents
Z413488
Sigma-Aldrich
Aldrich® Claisen adapter
With thermometer joint, without drip-tip, bottom and side joint: ST/NS 19/22
Synonym(s):
distilling adapter, glass adapter, glass adapters
About This Item
UNSPSC Code:
41121815
joint
bottom and side joint: ST/NS19/22
top joint ST/NS10/18
General description
A 50mm immersion length thermometer is recommended.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)