Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z413224

Sigma-Aldrich

Aldrich® Vigreux column

Unjacketed, effective L 50 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812

joint

joint ST/NS24/40

I.D. × O.D.

19 mm × 22 mm

effective L

50 mm

Looking for similar products? Visit Product Comparison Guide

General description

Vigreux indents run the entire length of the column and act as the packing for the column.
Jacketed version is pictured.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service