Z412139
BD Precisionglide® syringe needles
gauge 22, L 1 in.
Synonym(s):
cannula
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
Recommended Products
packaging
pkg of 100 ea
manufacturer/tradename
Becton-Dickinson & Co. 305155
L
1 in.
gauge
22
color
black
Looking for similar products? Visit Product Comparison Guide
General description
- Fits any Luer lock, slip tip, or eccentric tip syringe
- SS Microlance regular-bevel point
- Siliconized to reduce resistance
- PP Luer hub color coded to needle gauge
- Packaged as individual fiber-free, peel-apart packages in strips of five sterile needles
- A sheath protects point and cannula
- All packages carry lot control numbers to insure sterility
- NoKor needles have scalpel like tip
Legal Information
Precisionglide is a registered trademark of Becton-Dickinson & Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service