Key Documents
Z411507
Sigma-Aldrich
150 and More Basic NMR Experiments, 2nd Ed.
About This Item
ISBN-10:
3527295127
ISBN-13:
978-352729512-8
UNSPSC Code:
55101509
publication info
S. Braun, et al., John Wiley & Sons, 1998, 610 pp., soft cover
manufacturer/tradename
Wiley
General description
This edition of this highly successful book is expanded by another 50 important NMR experiments dedicated to the latest developments. The expansions mainly concern the fields of chemical applications, of very recent gradient selected experiments now arranged in two chapters (1D and 2D), and of the basics of solid-state NMR in a completely new chapter. Again, all experiments from basic pulse determinations to advanced 2D and some 3D techniques have been performed especially for this book. The figures are taken directly from the spectrometer output and the reader can therefore rely on the results presented. Written by leading experts, it is a thorough guide through the maze of modern NMR tools. Being a textbook as well as a reference and work book for the laboratory, this book is a must for every scientist applying NMR as well as for students preparing for their lab courses.
Table of Contents
The NMR Spectrometer.
Determination of the Pulse-Duration.
Routine NMR Spectroscopy and Standard Tests.
Decoupling Techniques.
Dynamic NMR Spectroscopy.
ID Multiple Sequences.
NMR Spectroscopy with Selective Pulses.
Auxiliary Reagents, Quantitative Determinations, and Reaction Mechanisms.
1D Heteronuclear NMR Spectroscopy.
The Second Dimension.
1D NMR Spectroscopy with Field Gradients.
2D NMR Spectroscopy with Field Gradients.
The Third Dimension.
Solid-State NMR Spectroscopy.
Appendices.
Glossary and Index.
Choose from one of the most recent versions:
Certificates of Analysis (COA)