Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z409456

Sigma-Aldrich

Weighing funnel

size L, borosilicate glass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.71

material

borosilicate glass

size

L

packaging

pkg of 6 ea

funnel I.D.

6.5 cm

neck O.D.

1.9 cm

Looking for similar products? Visit Product Comparison Guide

General description

  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service