Key Documents
Z403164
Büchi® vertical condenser, for Rotavapor® evaporators
for R200/205 and R210/215, glass
About This Item
UNSPSC Code:
41100000
material
glass
manufacturer/tradename
Büchi 040601
General description
Comes with KD-22 vacuum seal.
Condensers for R200/205 and R210/215 come with hose barbs, drain sleeve and 3m silicone hose. Replacement condensers do not include stopcocks.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)