Key Documents
Z402559
Best® Nitri Solve gloves
Flock-lined, L × thickness 13 in. × 15 mil, size XL
About This Item
UNSPSC Code:
46181504
size
XL
feature
reusable
packaging
pkg of 1 pair
manufacturer/tradename
Best Manufacturing 730-10
L × thickness
13 in. × 15 mil
General description
• Unsupported
• Offers good dexterity
• Non-slip grip
• Cut and puncture resistant
• Excellent for handling most aromatic, petroleum, and chlorinated solvents, as well as acids and caustics
Legal Information
Best is a registered trademark of Showa Glove Co.
Nitri Solve is a trademark of Best Manufacturing Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)