Key Documents
Z401722
MSA air-purifying Super Cartridges
GME/P100 type, for Ultra-Twin®respirator
About This Item
UNSPSC Code:
46182002
packaging
pkg of 6 ea
manufacturer/tradename
MSA 815182
availability
not available in EU
General description
These combination Super Cartridges provide simultaneous protection against multiple hazards and simplifies the selection process and training required for use.
Cartridge type NIOSH-approved for:
GME
Organic vapors, sulfur dioxide, chlorine, chlorine dioxide, hydrogen chloride, hydrogen
sulfide (escape only), hydrogen fluoride gases, ammonia, methylamine, formaldehyde
GME/P100 All of the above plus: paint, lacquer, enamels, pesticides (GMP), dusts*, mists*,
fumes*, asbestos containing dusts and mists, radionuclides
*Having a TWA < 0.05mg/m3 (i.e., toxic dusts)
Legal Information
Ultra-Twin is a registered trademark of Mine Safety Appliances Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)