Z401374
KDS dual-syringe pump series 200
AC/DC input 110 V AC
Synonym(s):
dosing pump, infusion pump, pump infusion
About This Item
Recommended Products
feature
CSA approved
1 channel
manufacturer/tradename
KD Scientific 780200-110V
availability
not available in EU
L × W × H
11 in. × 9 in. × 5 1/2 in.
volume
0.01-140 mL
weight
9 lb
suitability
suitable for MS calibration
suitable for aerosol spray
suitable for chemical reactor injection
suitable for electrospinning
suitable for infusion
General description
Easy setup:
1. Select syringe size
2. Enter volume to be dispensed
3. Enter flow rate, then press start button
Series 200 infusion pumps have all the features of Series 100 pumps plus:
- Holds two syringes, sizes 10 μL to 140 mL each
- Settings can be reviewed or changed during operation
- Optical encoder stall detection
- Choice of unit selection
- RS-232C interface for computerizing up to 100 pumps
- TTL interface for foot switch, timer, relay control
- Outputs for run indicator, valve control
Choose from one of the most recent versions:
Certificates of Analysis (COA)