Key Documents
Z314242
Grinding balls for Retsch MM200 mixer mill
stainless steel, diam. 9-10 mm, bulk density 7.7 g/mL
About This Item
UNSPSC Code:
41101708
material
stainless steel
manufacturer/tradename
Retsch
availability
not available in USA and Canada
diam.
9-10 mm
bulk density
7.7 g/mL
General description
Recommend use grinding jars of same material where possible
Choose from one of the most recent versions:
Certificates of Analysis (COA)