Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z309532

Quickfit® flat-bottom boiling flasks

capacity 4,000 mL, joint: ST/NS 45/40

Synonym(s):

FF4L/7S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804

material

borosilicate glass
flat bottom flask
short

joint

joint: ST/NS45/40

sterility

non-sterile

feature

closure type Standard Taper Joint

packaging

case of 1

manufacturer/tradename

Quickfit FF 2L/7S

availability

not available in USA and Canada

capacity

4,000 mL

Looking for similar products? Visit Product Comparison Guide

General description

With short neck and ground-glass socket. Not suitable for vacuum use.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service