Key Documents
Z309451
Quickfit® flat-bottom boiling flasks
capacity 500 mL, joint: ST/NS 34/35
Synonym(s):
FF500/5S
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass flask
flat bottom
single-neck
joint
joint: ST/NS34/35
packaging
pack of 4 ea
manufacturer/tradename
Quickfit FF 500/5S
availability
not available in USA
capacity
500 mL
General description
With short neck and ground-glass socket. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)