Key Documents
Z304964
Frigimat® economy dry ice makers
European connections
About This Item
UNSPSC Code:
41103000
manufacturer/tradename
Bel-Art
General description
A 6ft nickel plated copper connecting tube with standard European thread for attachment to CO2 cylinder is included.
Produces solid 1lb cylindrical block 9 × 10 cm of dry ice from a cylinder of liquid CO2. One CO2 cylinder makes 8 to 9 blocks of dry ice. Epoxy-coated steel frame, brass valve, PE dry ice chamber.
This product is a dry ice maker that produces a solid 454 gm (16 oz) block of dry ice in one to two minutes. Each chamber of liquid CO2 will make 8 or 9 blocks of dry ice measuring 92 mm x 102 mm long (3-5/8"x 4"). A polyethylene ice chamber is held in place by a metal clamping mechanism and a porous polyethylene diffusion plate vents excess gas.
The unit also features a built-in gas pressure gauge and relief valve. The epoxy-coated steel frame has four non-skid rubber feet to prevent sliding. Overall dimensions are 37x14x15 cm (14-3/4"x 5-1/2"x 6") high with a shipping weight of 3.9 kg (8-1/2 lbs).
Includes a 1.8 m (6 ft) nickel plated copper connecting tube with standard GCA 320 (.830"-14) thread couplings for easy attachment to a LCO2 cylinder.
Legal Information
Frigimat is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)