Key Documents
Z304859
Quickfit® drying tube
joint: ST/NS 29/32
Synonym(s):
MF23/4
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
material
colorless tube
glass tube
joint
joint: ST/NS29/32
sterility
non-sterile
feature
graduations: no
neck thread PUSH FIT (29/32 cone size)
manufacturer/tradename
Quickfit MF 23/4
availability
not available in USA and Canada
binding type
non-treated surface
General description
Angled. Retain desiccant with plug of glass wool.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)