Key Documents
Z304190
Quickfit® column packing supports
fits ST/NS: 19/26 female joint
Synonym(s):
EX13/12
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
manufacturer/tradename
Quickfit® EX 13/12
availability
not available in ROW (Available only in EU countries), USA and Canada
not available in USA and Canada
diam.
16 mm
General description
These spiral packing supports will hold 4 mm or larger Fenske helices. They may be used with larger socket sizes than those shown below providing there is a suitable constriction at or above the cone at the bottom of the column.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)