Key Documents
Z303410
Plastic hose connectors for Quickfit® water-cooled condenser
Synonym(s):
4510/02
About This Item
UNSPSC Code:
41103025
NACRES:
NB.35
Product Z303410 is not currently sold in your country. Contact Technical Service
packaging
pkg of 10 ea
manufacturer/tradename
Quickfit
availability
available only in EU
General description
These screwthread hose connectors are only for use with the QUICKFIT® and PYREX® glassware products. They do not fit DURAN® glass threads.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
PYREX is a registered trademark of Corning, Inc.
QUICKFIT is a registered trademark of DWK Life Sciences
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)