Key Documents
Z302252
Tyvek® Pro.Tech coveralls
size XXL
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
size
XXL
manufacturer/tradename
Tyvek
availability
not available in USA and Canada
General description
With integral hood, full length zipper and elasticated wrist and ankle holes. Zipper has covering flap and seams are taped for extra protection. Provides CE Type 5 and Type 6 (particle and limited splash) protection.
Legal Information
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)