Key Documents
Z301698
Marigold® Suretech vinyl gloves
size M, standard length
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
size
M
feature
CE compliant
standard length
packaging
pkg of 1000 ea
manufacturer/tradename
Marigold
availability
not available in USA and Canada
L × thickness
240 mm × 0.26 mm
General description
Available in standard or 25% longer length, these ambidextrous gloves are ideal for use in laboratory, pharmaceutical or electronic environments. Double bagged, powder-free and with low Chloride, Sulfur and Sodium ion levels, they combine minimum particulate contamination with good sensitivity, grip and tear resistance.
Legal Information
Marigold is a registered trademark of Comasec SAS
Choose from one of the most recent versions:
Certificates of Analysis (COA)