Key Documents
Z301043
Quickfit® Reduction Adapters
top female joint: ST/NS 34/35, bottom male joint: ST/NS 40/38
Synonym(s):
DA56
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
female joint: ST/NS top34/35
male joint: ST/NS bottom40/38
manufacturer/tradename
Quickfit DA 56
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)