Key Documents
Z292117
Supelco
Decapper
O.D. 8 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
O.D.
8 mm
General description
to remove aluminum seals from bottles and vials
Application
For aluminum seal vials
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Crimpers Hand crimper, for use with 20mm seals, pkg of 1 ea
33280-U
Crimpers