Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z290238

Supelco

Model 1000 oxygen trap

connections size 1/4 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NB.21

connections size

1/4 in.

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

High-pressure, nickel-plated steel cylinder filled with a high capacity oxygen adsorbent. Low initial cost and high capacity gives months of use and reduces down time. Exceptional efficiency typically results in less than 30 ppb oxygen at the outlet.

Oxygen traps protect the liquid phase in HPLC columns from oxidation, and are particularly useful for capillary columns.

Contains 500 g of adsorbent, capable of removing over 1 L of oxygen at standard temperature and pressure.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Self-heat. 1 - Skin Irrit. 2 - STOT RE 2 Inhalation

target_organs

Lungs

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service