Key Documents
Z288330
KNF Laboport® vacuum system components
System baseplate for 34 L/min pump
About This Item
UNSPSC Code:
41105101
NACRES:
NB.79
description
Baseplate for pump and vacuum controller C only.
feature
CE compliant
manufacturer/tradename
KNF
General description
Modular equipment with plug-in connectors for building systems that will support many laboratory vacuum requirements. Components do not include 3/8 in. I.D. vacuum tubing (see the Tubing Section) or tool kit, available separately. All components are sold separately.
Supports the modular components and vacuum pump. Baseplates are sized to fit the pump.
Legal Information
Laboport is a registered trademark of KNF Neuberger GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)