Key Documents
Z286680
Positioning rod for WILMAD® 5 mm microcell assemblies
helps place the microcell in the NMR tube
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
Select a Size
1 EA
$44.10
$44.10
Available to ship onApril 29, 2025Details
Select a Size
Change View
1 EA
$44.10
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
$44.10
Available to ship onApril 29, 2025Details
packaging
pack of 1 ea
manufacturer/tradename
Wilmad 529-C
General description
The positioning rod helps place the microcell precisely in the Wilmad sample NMR tube. Remove the positioning rod prior to recording the spectrum.
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)