Key Documents
Z286664
Microcells for WILMAD® 5 mm microcell assemblies
spherical, volume 18 μL
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
packaging
pack of 10 ea
manufacturer/tradename
Wilmad 529-A
volume
18 μL
General description
Microcells may be used interchangeably in the PTFE holder of the Microcell Assembly and can give the flexibility of taking NMR spectra of small samples.
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)