Key Documents
Z286192
Sigma-Aldrich
Aldrich® pocket NMR tube carrier
assorted colors, pkg of 3 ea
About This Item
UNSPSC Code:
41100000
packaging
pkg of 3 ea
color
assorted colors
General description
One of each color carrier; red, white, and blue.
Unbreakable NMR tube carrier fits into labcoat pocket like a pen. Provides protection for sample and lab personnel when transporting samples. Has a snap-on cap and a shirt clip that secures carrier in pocket. Holds one 5 mm diam. × 7 in. L NMR tube.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)