Key Documents
Z285110
Glas-Col® Series O extension support
for flask size, 3000 mL
About This Item
UNSPSC Code:
41102405
NACRES:
NB.76
material
steel
feature
for flask size 3000 mL
manufacturer/tradename
Glas-Col 100E0545
General description
The steel support rings with fabric strap basket hold spherical and hemispherical mantles. Attach to lattice systems or ring stands.
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)