Key Documents
Z284106
Spinring® magnetic stirring bar
size 1/2 in. × 5/16 in.
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
PTFE-coated
octagonal PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37140-0005
L
12.7 mm
diam.
8 (19mm ring O.D) mm
size
1/2 in. × 5/16 in.
General description
Bars with a removable stabilizing ring to keep bar centered.
Legal Information
Spinring is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)