Key Documents
Z283932
Sigma-Aldrich
Spinwedge® magnetic stirring bar
size 3/8 in. × 1/2 in.
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
PTFE-coated (wedge shaped)
PTFE-coated
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art F37123-0001
L
12.7 mm
diam.
9.5 mm
size
3/8 in. × 1/2 in.
General description
  • Spinwedge® magnetic stirring bars are ideal for churning sediment or dissolving salts for thorough dispersion.
  • The wedge shape design provides strong turbulence at low speeds.
  • The chemically resistant bars feature a sharply angled broad face for rigorous mixing of viscous solutions.
  • Alnico magnet encapsulated in FDA Grade PTFE (polytetrafluoroethylene) coating.
  • Autoclavable.
Bars designed to plough up sediment and provide strong mixing action.
Legal Information
Spinwedge is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)