Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z281689

Stockhausen skin protection creams

DuroPro

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103200

General description

Designed to protect skin from all kinds of professional hazards, these products are applied before starting work. All creams are supplied in 100 mL tubes.
Water repellant for acids, lyes, salts, cleaning agents and disinfectants. Also for cement, lime, and fertilizer. Good for rough, dry, or chapped hands.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service