Key Documents
Z281573
Sigma-Aldrich
Oakton® thermometer/timer
AAA battery included
About This Item
UNSPSC Code:
41122411
NACRES:
NB.35
manufacturer/tradename
Oakton
General description
  • Audible alarm for temp. and time
  • Temp.: 0 to 200 °C (32 to 392 °F)
  • Time: Counts down from 23h, 59min., 59s
Large LCD display with easily set temp. and time set points. Select from °F or °C.
Measure temperature using a 6 1/2 in. long stainless steel probe with a 3 1/2 ft long lead. AAA battery included.
Legal Information
Oakton is a registered trademark of Cole-Parmer Instrument Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)