Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z281565

Sigma-Aldrich

Micro-90® concentrated cleaning solution

volume 4 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800

technique(s)

HPLC: suitable
LC/MS: suitable

volume

4 L
40000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Cleaning methods include CIP, Manual and Immersion Washing, Mild-Agitation, Ultrasonic. Typical pH (neat) 9.0 to 9.9. Five-year shelf life. A mild yet powerful multi-purpose cleaner that is effective in both industrial and critical cleaning applications. Its unique formulation of chelants, ionic, and non-ionic ingredients produces a variety of cleaning actions that include lifting, dispersing, emulsifying, sequestering, suspending and decomposing. Excellent for cleaning soils include oil, grease, wax, tar, flux, particulates, and hard water stains. Outstanding for cleaning labware, ceramics, stainless steel, and defouling filter membranes.
Concentrate: dilute to 2% Micro in water.
May etch aluminum and zinc.

Legal Information

Micro-90 is a registered trademark of International Products

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2

target_organs

Respiratory Tract

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service